missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Chloromethyl)-3-(4-chlorophenyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Formula Weight | 229.07 |
Percent Purity | 95% |
Quantity | 10 g |
Chemical Name or Material | 5-(Chloromethyl)-3-(4-chlorophenyl)-1, 2, 4-oxadiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'