missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-2-thien-2-ylpyrimidine, 97%, Thermo Scientific™

Product Code. 10401884
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 926921-78-0
Molecular Formula C9H7ClN2S
Molecular Weight (g/mol) 210.68
MDL Number MFCD09879950
InChI Key HBGUDWOIHDHRCQ-UHFFFAOYSA-N
Synonym 5-chloromethyl-2-thien-2-ylpyrimidine, 5-chloromethyl-2-thiophen-2-yl pyrimidine, 2-5-chloromethyl pyrimidin-2-yl thiophene, 5-chloromethyl-2-thiophen-2-ylpyrimidine
PubChem CID 24229732
IUPAC Name 5-(chloromethyl)-2-thiophen-2-ylpyrimidine
SMILES ClCC1=CN=C(N=C1)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 210.69
Percent Purity 97%
Chemical Name or Material 5-(Chloromethyl)-2-thien-2-ylpyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'