missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Chloromethyl)-2-phenylpyrimidine, 97%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 1g
Specifications
CAS | 886531-63-1 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C11H9ClN2 |
MDL Number | MFCD09879948 |
Quantity | 1 g |
Synonym | 5-chloromethyl-2-phenylpyrimidine, 5-chloromethyl pyrimidin-2-yl benzene, 5-chloromethyl-2-phenyl-1,3-diazine, pyrimidine, 5-chloromethyl-2-phenyl, pyrimidine, 5-chloromethyl-2-phenyl-9ci |
InChI Key | VJINEZQXTOUKNJ-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'