Learn More
5-(Chloromethyl)-1-methyl-3-thien-2-yl-1H-pyrazole, ≥97%, Maybridge™
Brand: Maybridge CC47224CB
Code : 31
Additional Details : CAS Number : 876316-96-0 Weight : 0.05025kg Transport : UN number : 3261 Chem class : 8 Pack group : III
Quantity | 250mg |
---|
Chemical Identifiers
876316-96-0 | |
212.695 | |
GQMRXNSYTBKJTB-UHFFFAOYSA-N | |
18525812 | |
CN1C(=CC(=N1)C2=CC=CS2)CCl |
C9H9ClN2S | |
MFCD08435886 | |
5-chloromethyl-1-methyl-3-thien-2-yl-1h-pyrazole, 5-chloromethyl-1-methyl-3-thiophen-2-yl pyrazole, 1h-pyrazole,5-chloromethyl-1-methyl-3-2-thienyl, 2-5-chloromethyl-1-methylpyrazol-3-yl thiophene, 5-chloromethyl-1-methyl-3-thiophen-2-yl-1h-pyrazole | |
5-(chloromethyl)-1-methyl-3-thiophen-2-ylpyrazole |
Specifications
5-(Chloromethyl)-1-methyl-3-thien-2-yl-1H-pyrazole | |
876316-96-0 | |
100.0 | |
C9H9ClN2S | |
5-chloromethyl-1-methyl-3-thien-2-yl-1h-pyrazole, 5-chloromethyl-1-methyl-3-thiophen-2-yl pyrazole, 1h-pyrazole,5-chloromethyl-1-methyl-3-2-thienyl, 2-5-chloromethyl-1-methylpyrazol-3-yl thiophene, 5-chloromethyl-1-methyl-3-thiophen-2-yl-1h-pyrazole | |
CN1C(=CC(=N1)C2=CC=CS2)CCl | |
212.695 | |
212.7 |
250mg | |
96.5 | |
97% | |
MFCD08435886 | |
GQMRXNSYTBKJTB-UHFFFAOYSA-N | |
5-(chloromethyl)-1-methyl-3-thiophen-2-ylpyrazole | |
18525812 | |
97% |