missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloroindole, 98%

Product Code. p-7021311
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410134 1 g 1g
11420134 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410134 Supplier Alfa Aesar™ Supplier No. A13592.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
5-Chloroindole has been used in the synthesis of 5-chloro-3-indole-N,N- dimethylglyoxalamide and 5-chloro-N,N-dimethyltryptamine. It may be used in the synthesis of dyestuffs in the presence of biocatalysts. 5-Chloroindole can be synthesized by using 3-chlorobenzaldehyde as starting reagent

Solubility
Soluble in alcohol.

Notes
Store at room temperature. Incompatible with acids, reducing agents and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17422-32-1
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00005672
InChI Key MYTGFBZJLDLWQG-UHFFFAOYSA-N
Synonym 5-chloroindole, 1h-indole, 5-chloro, 5-chloro indole, 1h-indole, 5-chloro-9ci, 5-chloroindol, 5-chloro-indole, 3fue, zlchem 215, 5-chlor-1h-indole, indole, 5-chloro
PubChem CID 87110
IUPAC Name 5-chloro-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1Cl

Specifications

Melting Point 69°C to 73°C
Quantity 1 g
Beilstein 2651
Formula Weight 151.6
Percent Purity 98%
Chemical Name or Material 5-Chloroindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.