missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate, 95%, Thermo Scientific™

Product Code. 15488975
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488975 250 mg 250mg
15478975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488975 Supplier Thermo Scientific Alfa Aesar Supplier No. H35034.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000017-93-5
Molecular Formula C8H5ClN2O2
Molecular Weight (g/mol) 196.59
MDL Number MFCD09836168
InChI Key TVUHAHMCORHJBN-UHFFFAOYSA-N
Synonym 5-chloroimidazo 1,2-a pyridine-2-carboxylic acid, 5-chloro-imidazo 1,2-a pyridine-2-carboxylic acid, 5-chloroimidazo 1,2-a pyridine-2-carboxylic acid hydrate, 2-carboxy-5-chloroimidazo 1,2-a pyridine, 5-chloroimidazo 1,2-a pyridine-2-carboxylicacid, imidazo 1,2-a pyridine-2-carboxylicacid,5-chloro, imidazo 1,2-a pyridine-2-carboxylic acid, 5-chloro, 5-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid
PubChem CID 26369183
IUPAC Name 5-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
SMILES OC(=O)C1=CN2C(C=CC=C2Cl)=N1

Specifications

Melting Point 140°C to 142°C
Quantity 250 mg
Formula Weight 196.59 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 5-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.