missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzo[b]thiophene, 97%, Thermo Scientific Chemicals

Product Code. 15488308
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20532-33-6
Molecular Formula C8H5ClS
Molecular Weight (g/mol) 168.638
MDL Number MFCD00055623
InChI Key SNYURIHMNFPQFL-UHFFFAOYSA-N
Synonym 5-chlorobenzothiophene, 5-chlorobenzo b thiophene, 5-chloro-benzo b thiophene, 5-chloro benzothiophene, benzo b thiophene, 5-chloro, zlchem 66, 5-chlorothionaphthene, 5-chloranyl-1-benzothiophene, benzo b thiophene,5-chloro, ksc544i5d
PubChem CID 11309754
IUPAC Name 5-chloro-1-benzothiophene
SMILES C1=CC2=C(C=CS2)C=C1Cl

Specifications

Color White
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 168.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Chlorobenzo[b]thiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'