missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroanthranilic Acid, TRC

Product Code. 30361352
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361352 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361352 Supplier Toronto Research Chemicals Supplier No. TRCC36440025G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloroanthranilic Acid
CAS 635-21-2
Molecular Formula C7H6ClNO2
Quantity 25 g
Synonym 2-Amino-5-chlorobenzoic Acid, 2-Carboxy-4-chloroaniline, 5-Chloro-2-aminobenzoic Acid, NSC 404157
InChI Formula InChI=1S/C7H6ClNO2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H,9H2,(H,10,11)
SMILES Nc1ccc(Cl)cc1C(=O)O
IUPAC Name 2-amino-5-chlorobenzoic acid
Molecular Weight (g/mol) 171.58
Formula Weight 171.0087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.