missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-m-phenylenediamine, 97%

Product Code. 11332536
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332536 5 g 5g
11342536 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332536 Supplier Thermo Scientific Alfa Aesar Supplier No. L06485.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33786-89-9 | C6H7ClN2 | 142.586 g/mol

5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33786-89-9
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00014775
InChI Key VZNUCJOYPXKLTA-UHFFFAOYSA-N
Synonym 5-chloro-m-phenylenediamine, 3,5-diaminochlorobenzene, 5-chloro-1,3-benzenediamine, 1,3-benzenediamine, 5-chloro, 5-chloro-1,3-phenylenediamine, ccris 2886, 5-amino-3-chloroaniline, acmc-20am42, 2-13-00-00029 beilstein handbook reference, 5-chloro-1,3-diaminobenzene
PubChem CID 73335
IUPAC Name 5-chlorobenzene-1,3-diamine
SMILES C1=C(C=C(C=C1N)Cl)N

Specifications

Melting Point 99°C to 103°C
Quantity 5 g
UN Number UN1673
Beilstein 3029854
Solubility Information Slightly soluble in water.
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-m-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.