missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzimidazole (Triclabendazole Sulfoxide), Mikromol™

Product Code. 17960005
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960005 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17960005 Supplier Mikromol Supplier No. MM0313.010025
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzimidazole (Triclabendazole Sulfoxide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triclabendazole sulfoxide
CAS 100648-13-3
Analyte or Component Names Triclabendazole sulfoxide
Shipping Condition Room Temperature
Molecular Formula C14 H9 Cl3 N2 O2 S
Certifications/Compliance ISO 9001
Synonym 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzimidazole, 1H-Benzimidazole, 5-chloro-6-(2,3-dichlorophenoxy)-2-(methylsulfinyl)- (9CI), 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzimidazole, 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzimidazole, CGA 110752, Triclabendazole Sulfoxide, Triclabendazole Related Compound A, Triclabendazole Impurity A
InChI Formula InChI=1S/C14H9Cl3N2O2S/c1-22(20)14-18-9-5-8(16)12(6-10(9)19-14)21-11-4-2-3-7(15)13(11)17/h2-6H,1H3,(H,18,19)
SMILES CS(=O)c1nc2cc(Cl)c(Oc3cccc(Cl)c3Cl)cc2[nH]1
IUPAC Name 5-chloro-6-(2,3-dichlorophenoxy)-2-methylsulfinyl-1H-benzimidazole
Molecular Weight (g/mol) 375.66
Quantity 25 mg
Formula Weight 373.945 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.