missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-4-methoxythiophene-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10160732
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10160732 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10160732 Supplier Thermo Scientific Maybridge Supplier No. KM00452CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 133659-14-0
Molecular Formula C6H5ClO3S
Molecular Weight (g/mol) 192.613
MDL Number MFCD00067999
InChI Key CEOFOUVVEMMYDW-UHFFFAOYSA-N
Synonym 3-thiophenecarboxylicacid, 5-chloro-4-methoxy, 5-chloro-4-methoxy-3-thiophenecarboxylic acid, maybridge4_002389, acmc-1c4c3, 5-chloro-4-methoxythiophene-3-carboxylicacid, 3-thiophenecarboxylicacid,5-chloro-4-methoxy, 5-chloranyl-4-methoxy-thiophene-3-carboxylic acid
PubChem CID 2778382
IUPAC Name 5-chloro-4-methoxythiophene-3-carboxylic acid
SMILES COC1=C(SC=C1C(=O)O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 192.62
Percent Purity 97%
Chemical Name or Material 5-Chloro-4-methoxythiophene-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.