missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-3-methylbenzo[b]thiophene, 97%, Thermo Scientific™

Product Code. 10232312
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10232312 50 g 50g
10762873 10 g 10g
10118744 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10232312 Supplier Thermo Scientific Maybridge Supplier No. MWP00002EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 19404-18-3
Molecular Formula C9H7ClS
Molecular Weight (g/mol) 182.665
MDL Number MFCD00052506
InChI Key UWDQVEPXORTQFO-UHFFFAOYSA-N
Synonym 5-chloro-3-methylbenzo b thiophene, 5-chloro-3-methylthianaphthene, 5-chloro-3-methyl benzo b thiophene, 5-chloro-3-methylbenzothiophene, 4-methylthio phenol, zlchem 217, pubchem7372, acmc-209exr, maybridge1_005366
PubChem CID 317317
IUPAC Name 5-chloro-3-methyl-1-benzothiophene
SMILES CC1=CSC2=C1C=C(C=C2)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 50 g
Formula Weight 182.67
Percent Purity 97%
Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.