missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10580922
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10580922 1 g 1g
11396905 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10580922 Supplier Thermo Scientific Maybridge Supplier No. MWP00006DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 166964-33-6
Molecular Formula C9H6Cl2O2S2
Molecular Weight (g/mol) 281.17
MDL Number MFCD00173797
InChI Key JLTUANWNGKWRQO-UHFFFAOYSA-N
Synonym 5-chloro-3-methylbenzo b thiophene-2-sulfonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulphonyl chloride, 5-chloro-3-methylbenzo b thiophene-2-sulfonyl, 5-chloro-3-methylbenzo b thiophene-2-sulphonylchloride, 5-chloro-3-methyl-benzo b thiophene-2-sulfonyl chloride, benzo b thiophene-2-sulfonylchloride, 5-chloro-3-methyl, zlchem 600, pubchem5100, buttpark 21\07-05, 5-chloro-3-methyl-benzothiophene-2-sulfonyl chloride
PubChem CID 2735756
IUPAC Name 5-chloro-3-methyl-1-benzothiophene-2-sulfonyl chloride
SMILES CC1=C(SC2=CC=C(Cl)C=C12)S(Cl)(=O)=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 281.18
Percent Purity 97%
Chemical Name or Material 5-Chloro-3-methylbenzo[b]thiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.