missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-methyl-4-nitro-1H-pyrazole, TRC

Product Code. 30365930
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. Quantity unitSize
30365930 10 g 10g
30351715 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 30365930

Brand: Toronto Research Chemicals TRCC36911510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6814-58-0
Formula Weight 160.9992
InChI Formula InChI=1S/C4H4ClN3O2/c1-2-3(8(9)10)4(5)7-6-2/h1H3,(H,6,7)
IUPAC Name 5-chloro-3-methyl-4-nitro-1H-pyrazole
Molecular Formula C4 H4 Cl N3 O2
Molecular Weight (g/mol) 161.546
SMILES Cc1n[nH]c(Cl)c1[N+](=O)[O-]
Synonym Pyrazachlor, Pyrazole, 3(or 5)-chloro-5(or 3)-methyl-4-nitro- (7CI), 3-Chloro-5-methyl-4-nitro-1H-pyrazole, 3-Chloro-4-nitro-5-methyl-1H-pyrazole, 5-Chloro-3-methyl-4-nitro-1H-pyrazole, ABG 3030, CMN-Pyrazole, Release, 1H-Pyrazole, 3-chloro-5-methyl-4-nitro-
Chemical Name or Material Pyrazachlor

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.