missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-methyl-1-benzothiophene-2-carboxylic acid, Technical Grade, Thermo Scientific™

Product Code. 10641683
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10641683

Brand: Thermo Scientific Maybridge MO00546DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50451-84-8
Molecular Formula C10H7ClO2S
Molecular Weight (g/mol) 226.67
MDL Number MFCD03407361
InChI Key KEYCXZNZFABITO-UHFFFAOYSA-N
Synonym 5-chloro-3-methylbenzo b thiophene-2-carboxylic acid, 5-chloro-3-methyl-1-benzothiophene-2-carboxylic ac, 5-chloro-3-methyl-benzo b thiophene-2-carboxylic acid, 5-chloro3-methyl-benzo b thiophene-2-carboxylic acid, 5-chloro-3-methyl-1-benzo b thiophene-2-carboxylic acid
PubChem CID 2794614
IUPAC Name 5-chloro-3-methyl-1-benzothiophene-2-carboxylic acid
SMILES CC1=C(SC2=CC=C(Cl)C=C12)C(O)=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 226.68
Percent Purity 95%
Chemical Name or Material 5-Chloro-3-methyl-1-benzothiophene-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.