missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-hydroxypyridine, 99%

Product Code. 11321957
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321957 25 g 25g
11311957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321957 Supplier Thermo Scientific Alfa Aesar Supplier No. L07568.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74115-12-1 | C5H4ClNO | 129.543 g/mol

5-Chloro-3-hydroxypyridine is used in the preparation of 4-(5-chloropyridin-3-yloxy)isoquinoline

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-3-hydroxypyridine is used in the preparation of 4-(5-chloropyridin-3-yloxy)isoquinoline

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74115-12-1
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD00006379
InChI Key TUIDQYRZDZRHPQ-UHFFFAOYSA-N
Synonym 5-chloro-3-hydroxypyridine, 5-chloro-3-pyridinol, 3-chloro-5-hydroxypyridine, 3-pyridinol, 5-chloro, 5chloro-3-pyridinol, pubchem6628, 3-chloro-5-pyridinol, acmc-209otf, 5-chloranylpyridin-3-ol, 3chloro-5-hydroxypyridine
PubChem CID 582305
IUPAC Name 5-chloropyridin-3-ol
SMILES C1=C(C=NC=C1Cl)O

Specifications

Melting Point 159°C to 163°C
Quantity 25 g
Beilstein 1422550
Solubility Information Soluble in water.
Formula Weight 129.55
Percent Purity 99%
Chemical Name or Material 5-Chloro-3-hydroxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.