missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(trichloromethyl)benzimidazole, 95%, Thermo Scientific™

Product Code. 10397360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10397360 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10397360 Supplier Thermo Scientific Acros Supplier No. 185300050
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 3584-66-5
CAS Min % 94.0
CAS Max % 100.0
Melting Point 223°C to 229°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H4Cl4N2
MDL Number MFCD00005595
Quantity 5 g
Synonym 5-chloro-2-trichloromethyl benzimidazole, 5-chloro-2-trichloromethyl-1h-benzo d imidazole, 5-chloro-2-trichloromethyl-1h-benzimidazole, 6-chloro-2-trichloromethyl-1h-benzimidazole, 5-chloro-2-trichloromethyl-1h-1,3-benzodiazole, 5-chloro-2-trichloromethyl-3h-1,3-benzodiazole, acmc-20am4z, 5-chloro-2-trichloromethyl-1h-benzoimidazole
InChI Key SIZGSKQSWJIWFP-UHFFFAOYSA-N
SMILES ClC1=CC=C2N=C(NC2=C1)C(Cl)(Cl)Cl
IUPAC Name 6-chloro-2-(trichloromethyl)-1H-benzimidazole
Molecular Weight (g/mol) 269.93
PubChem CID 77134
Formula Weight 269.94
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Chloro-2-(trichloromethyl)benzimidazole, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Chloro-2-(trichloromethyl)benzimidazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.