missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-toluidine, TRC

Product Code. 30362327
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362327 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362327 Supplier Toronto Research Chemicals Supplier No. TRCC42160550G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2-methylaniline
CAS 95-79-4
Molecular Formula C7 H8 Cl N
Quantity 50 g
Synonym Fast Red KB Base (6CI), o-Toluidine, 5-chloro- (8CI), 5-Chloro-2-methylbenzenamine, 2-Amino-4-chlorotoluene, 2-Methyl-5-chloroaniline, 3-Chloro-6-methylaniline, 4-Chloro-2-aminotoluene, 5-Chloro-2-methylaniline, 5-Chloro-2-methylphenylamine, 5-Chloro-2-toluidine, 5-Chloro-o-toluidine, Acco Fast Red KB base, Ansibase Red KB, Azoene Fast Red KB base, Fast Red KB amine, Fast Red KB salt, Fast Red KB salt Supra, Fast Red KB-T Base, Fast Red KBS salt, Genazo Red KB soln, Hiltonil Fast Red KB base, Metrogen Red Former KB soln, NSC 7094, Naphthosol Fast Red KB base, Pharmazoid Red KB, Red KB base, Spectrolene Red KB, Stable Red KB base
InChI Formula InChI=1S/C7H8ClN/c1-5-2-3-6(8)4-7(5)9/h2-4H,9H2,1H3
SMILES Cc1ccc(Cl)cc1N
IUPAC Name 5-chloro-2-methylaniline
Molecular Weight (g/mol) 141.6
Formula Weight 141.0345
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.