missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-thiophenecarboxylic Acid, TRC

Product Code. 30134191
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134191 50 g 50g
30008783 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134191 Supplier Toronto Research Chemicals Supplier No. TRCC42050050G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24065-33-6
Molecular Formula C5 H3 Cl O2 S
Molecular Weight (g/mol) 162.59
Synonym 5-Chloro-2-thiophenecarboxylic Acid, 2-Chloro-5-carboxythiophene, 2-Chloro-5-thiophenecarboxylic acid, 5-Chloro-2-thiophenecarboxylic acid, 5-Chlorothiophen-2-carboxylic acid, NSC 14776
IUPAC Name 5-chlorothiophene-2-carboxylic acid
SMILES OC(=O)c1ccc(Cl)s1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H3ClO2S/c6-4-2-1-3(9-4)5(7)8/h1-2H,(H,7,8)
Formula Weight 161.9542
Chemical Name or Material 5-Chlorothiophene-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.