missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-thiophenecarboxaldehyde, 97%

Product Code. 10639384 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10639384 1 g Glass bottle 1g
10239740 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10639384 Supplier Thermo Scientific Acros Supplier No. 405080010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 7283-96-7 | C5H3ClOS | 146.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7283-96-7
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00047090
InChI Key VWYFITBWBRVBSW-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxaldehyde, 2-chloro-5-thiophenecarboxaldehyde, 5-chlorothiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 5-chloro, 5-chloro-2-formylthiophene, 2-chloro-5-formylthiophene, 5-chlorothiophene-2-aldehyde, 5-chloro-2-thiophenecarbaldehyde, 5-chloro-2-thiophene-carboxaldehyde, pubchem7742
PubChem CID 81700
IUPAC Name 5-chlorothiophene-2-carbaldehyde
SMILES ClC1=CC=C(S1)C=O

Spezifikation

CAS Min % 96.0
CAS Max % 100.0
Color Tan to White
Density 1.3760g/mL
Boiling Point 94°C (18.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6040 to 1.606
Quantity 1 g
Specific Gravity 1.376
Solubility Information Solubility in water: insoluble
Formula Weight 146.6
Percent Purity 97%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 5-Chloro-2-thiophenecarboxaldehyde, 97%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 5-Chloro-2-thiophenecarboxaldehyde
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.