missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-pyridinamine, TRC

Product Code. 17981455
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981455 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17981455 Supplier Toronto Research Chemicals Supplier No. TRCC3803401G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-98-6
Molecular Formula C5 H5 Cl N2
Molecular Weight (g/mol) 128.56
Synonym 2-Amino-5-chloropyridine, 2-Pyridinamine, 5-chloro- (9CI, ACI), 5-Chloro-2-pyridinamine (ACI), Pyridine, 2-amino-5-chloro- (6CI, 7CI, 8CI), (5-Chloropyridin-2-yl)amine, 5-Chloro-2-aminopyridine, 5-Chloro-2-pyridineamine, 5-Chloro-α-aminopyridine, NSC 26283, RP 26695
IUPAC Name 5-chloropyridin-2-amine
SMILES Nc1ccc(Cl)cn1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H5ClN2/c6-4-1-2-5(7)8-3-4/h1-3H,(H2,7,8)
Formula Weight 128.0141
Percent Purity >95%
Chemical Name or Material 2-Amino-5-chloropyridine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.