missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-pentanone ethylene ketal, 97%, Thermo Scientific Chemicals

Catalog Number p-90904
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5978-08-5
C7H13ClO2
164.63
MFCD00003217
OFERIRWCHSOJJT-UHFFFAOYSA-N
This item is not returnable. View return policy
5978-08-5
C7H13ClO2
164.63
MFCD00003217
OFERIRWCHSOJJT-UHFFFAOYSA-N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 73°C to 76°C (7.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4490 to 1.451
Quantity 5 g
Specific Gravity 1.09
Synonym 2-3-chloropropyl-2-methyl-1,3-dioxolane, 5-chloro-2-pentanone ethylene ketal, 2,2-ethylenedioxy-5-chloropentane, 1,3-dioxolane, 2-3-chloropropyl-2-methyl, 5-chloro-2-pentanoneethyleneketal, acmc-20am4f, oferirwchsojjt-uhfffaoysa, 1-chloro-4-pentanone ethylene ketal, 1, 2-3-chloropropyl-2-methyl, 5-chloro-2-pentanone ethylene acetal
SMILES CC1(OCCO1)CCCCl
IUPAC Name 2-(3-chloropropyl)-2-methyl-1,3-dioxolane
PubChem CID 80079
Formula Weight 164.63
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-pentanone ethylene ketal, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'