missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-pentanone, 94%

Product Code. p-7080590
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433993 25 g 25g
11443993 100 g 100g
11354659 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433993 Supplier Thermo Scientific Alfa Aesar Supplier No. A10269.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5891-21-4 | C5H9ClO | 120.576 g/mol

5-Chloro-2-pentanone used in the preparation of destructible surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-pentanone used in the preparation of destructible surfactants.

Solubility
Soluble in chloroform and methanol.

Notes
keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5891-21-4
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.576
MDL Number MFCD00001008
InChI Key XVRIEWDDMODMGA-UHFFFAOYSA-N
Synonym 5-chloro-2-pentanone, 2-pentanone, 5-chloro, 1-chloro-4-pentanone, 3-chloropropyl methyl ketone, 2-pentanone 5-chloro, 1-chloro-4-oxopentane, pubchem17207, 3-acetylpropyl chloride, 5-chloro-pentan-2-one, 5-chloranylpentan-2-one
PubChem CID 79993
IUPAC Name 5-chloropentan-2-one
SMILES CC(=O)CCCCl

Specifications

Boiling Point 63°C to 64°C (12 mmHg)
Flash Point 35°C (95°F)
Quantity 25 g
UN Number UN1224
Beilstein 1098490
Formula Weight 120.58
Percent Purity 94%
Chemical Name or Material 5-Chloro-2-pentanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.