missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-pentanone (>90%), TRC

Product Code. 30363052
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363052 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363052 Supplier Toronto Research Chemicals Supplier No. TRCC37509010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5891-21-4
Molecular Formula C5H9ClO
Quantity 10 g
Synonym 1-Chloro-4-pentanone, 3-Chloropropyl Methyl Ketone, NSC 60210, NSC 8427
InChI Formula InChI=1S/C5H9ClO/c1-5(7)3-2-4-6/h2-4H2,1H3
SMILES CC(=O)CCCCl
IUPAC Name 5-chloropentan-2-one
Molecular Weight (g/mol) 120.58
Formula Weight 120.0342
Chemical Name or Material 5-Chloro-2-pentanone
Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.