missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-pentanone, 90%

Product Code. 10076412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10076412 100 g Glass bottle 100g
10117303 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10076412 Supplier Thermo Scientific Acros Supplier No. 388291000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5891-21-4 | C5H9ClO | 120.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5891-21-4
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.58
MDL Number MFCD00001008
InChI Key XVRIEWDDMODMGA-UHFFFAOYSA-N
Synonym 5-chloro-2-pentanone, 2-pentanone, 5-chloro, 1-chloro-4-pentanone, 3-chloropropyl methyl ketone, 2-pentanone 5-chloro, 1-chloro-4-oxopentane, pubchem17207, 3-acetylpropyl chloride, 5-chloro-pentan-2-one, 5-chloranylpentan-2-one
PubChem CID 79993
IUPAC Name 5-chloropentan-2-one
SMILES CC(=O)CCCCl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Brown to Black
Density 1.0530g/mL
Boiling Point 172.0°C to 175.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3COCH3
Refractive Index 1.4370 to 1.4460
Quantity 100 g
Specific Gravity 1.053
Formula Weight 120.58
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-pentanone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Chloro-2-pentanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.