Learn More
5-chloro-2-nitro-4-(trifluoromethyl)aniline, 97%, Maybridge™
Brand: Maybridge CD04371CB
Code : 31
Additional Details : CAS Number : 35375-74-7 Weight : 0.05025kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
35375-74-7 | |
240.566 | |
AAMCWLIPRMIPBN-UHFFFAOYSA-N | |
2773813 | |
C1=C(C(=CC(=C1[N+](=O)[O-])N)Cl)C(F)(F)F |
C7H4ClF3N2O2 | |
MFCD01312277 | |
5-chloro-2-nitro-4-trifluoromethyl aniline, 4-amino-2-chloro-5-nitrobenzotrifluoride, 5-chloro-2-nitro-4-trifluoromethyl benzenamine, 3-chloro-6-nitro-4-trifluoromethyl aniline, 5-chloro-2-nitro-4-trifluoromethyl-phenylamine, 5-chloro-2-nitro-4-trifluoromethyl phenylamine, 5-chloro-4-trifluoromethyl-2-nitroaniline, benzenamine, 5-chloro-2-nitro-4-trifluoromethyl, 5-chloro-2-nitro-4-trifluoromethylphenyl amine, pubchem2803 | |
5-chloro-2-nitro-4-(trifluoromethyl)aniline |
Specifications
5-chloro-2-nitro-4-(trifluoromethyl)aniline | |
250mg | |
96.5 | |
97% | |
MFCD01312277 | |
AAMCWLIPRMIPBN-UHFFFAOYSA-N | |
5-chloro-2-nitro-4-(trifluoromethyl)aniline | |
2773813 | |
97% |
Amber Glass Bottle | |
35375-74-7 | |
100.0 | |
C7H4ClF3N2O2 | |
5-chloro-2-nitro-4-trifluoromethyl aniline, 4-amino-2-chloro-5-nitrobenzotrifluoride, 5-chloro-2-nitro-4-trifluoromethyl benzenamine, 3-chloro-6-nitro-4-trifluoromethyl aniline, 5-chloro-2-nitro-4-trifluoromethyl-phenylamine, 5-chloro-2-nitro-4-trifluoromethyl phenylamine, 5-chloro-4-trifluoromethyl-2-nitroaniline, benzenamine, 5-chloro-2-nitro-4-trifluoromethyl, 5-chloro-2-nitro-4-trifluoromethylphenyl amine, pubchem2803 | |
C1=C(C(=CC(=C1[N+](=O)[O-])N)Cl)C(F)(F)F | |
240.566 | |
240.57 |