missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Chloro-2-nitro-4-(trifluoromethyl)aniline, 95%

Product Code. 15448936
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448936 250 mg 250mg
15451669 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448936 Supplier Alfa Aesar™ Supplier No. H64456.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 35375-74-7
Molecular Formula C7H4ClF3N2O2
Molecular Weight (g/mol) 240.566
MDL Number MFCD01312277
InChI Key AAMCWLIPRMIPBN-UHFFFAOYSA-N
Synonym 5-chloro-2-nitro-4-trifluoromethyl aniline, 4-amino-2-chloro-5-nitrobenzotrifluoride, 5-chloro-2-nitro-4-trifluoromethyl benzenamine, 3-chloro-6-nitro-4-trifluoromethyl aniline, 5-chloro-2-nitro-4-trifluoromethyl-phenylamine, 5-chloro-2-nitro-4-trifluoromethyl phenylamine, 5-chloro-4-trifluoromethyl-2-nitroaniline, benzenamine, 5-chloro-2-nitro-4-trifluoromethyl, 5-chloro-2-nitro-4-trifluoromethylphenyl amine, pubchem2803
PubChem CID 2773813
IUPAC Name 5-chloro-2-nitro-4-(trifluoromethyl)aniline
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])N)Cl)C(F)(F)F

Specifications

Melting Point 112°C to 114°C
Color Yellow
Quantity 250 mg
UN Number UN2811
Formula Weight 240.57
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Chloro-2-nitro-4-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.