missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-(methylthio)pyrimidine-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10212872
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 61727-33-1
Molecular Formula C6H5ClN2O2S
Molecular Weight (g/mol) 204.628
MDL Number MFCD00173907
InChI Key SEPCYCDQJZTPHO-UHFFFAOYSA-N
Synonym 5-chloro-2-methylthio pyrimidine-4-carboxylic acid, 5-chloro-2-methylsulfanyl pyrimidine-4-carboxylic acid, 4-pyrimidinecarboxylic acid, 5-chloro-2-methylthio, 5-chloro-2-methylsulfanyl-pyrimidine-4-carboxylic acid, 4-carboxy-5-chloro-2-methylthio pyrimidine, 5-chloro-2-methylthio-4-pyrimidinecarboxylic acid, 5-chloro-2-methylthiopyrimidine-4-carboxylic acid, zlchem 91, oleosol fast red bl, oleosol fast red gl
PubChem CID 725711
IUPAC Name 5-chloro-2-methylsulfanylpyrimidine-4-carboxylic acid
SMILES CSC1=NC=C(C(=N1)C(=O)O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.64
Percent Purity 97%
Chemical Name or Material 5-chloro-2-(methylthio)pyrimidine-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'