missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylphenyl isothiocyanate, 97%, Thermo Scientific™

Catalog Number 11499576
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
19241-36-2
C8H6ClNS
183.653
MFCD00046801
VDBDGAPNWNWUSA-UHFFFAOYSA-N
5-chloro-2-methylphenyl isothiocyanate, isothiocyanic acid, 5-chloro-o-tolyl ester, 5-chloro-o-tolyl isothiocyanate, 5-chloro-2-methylphenylisothiocyanate, benzene,4-chloro-2-isothiocyanato-1-methyl, acmc-20am4h, 3-12-00-01913 beilstein handbook reference, 4-chloro-2-isothiocyanatotoluene, 5-chloro-2-methylbenzenisothiocyanate, 5-chloro-2-methylphenyl-isothiocyanate
29519
4-chloro-2-isothiocyanato-1-methylbenzene
CC1=C(C=C(C=C1)Cl)N=C=S
This item is not returnable. View return policy
19241-36-2
C8H6ClNS
183.653
MFCD00046801
VDBDGAPNWNWUSA-UHFFFAOYSA-N
5-chloro-2-methylphenyl isothiocyanate, isothiocyanic acid, 5-chloro-o-tolyl ester, 5-chloro-o-tolyl isothiocyanate, 5-chloro-2-methylphenylisothiocyanate, benzene,4-chloro-2-isothiocyanato-1-methyl, acmc-20am4h, 3-12-00-01913 beilstein handbook reference, 4-chloro-2-isothiocyanatotoluene, 5-chloro-2-methylbenzenisothiocyanate, 5-chloro-2-methylphenyl-isothiocyanate
29519
4-chloro-2-isothiocyanato-1-methylbenzene
CC1=C(C=C(C=C1)Cl)N=C=S

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
UN Number UN2922
Beilstein 2717659
Formula Weight 183.66
Percent Purity 97%
Density 1.248
Boiling Point 268°C to 272°C
Quantity 5 g
Odor Irritating
Refractive Index 1.646
Chemical Name or Material 5-Chloro-2-methylphenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'