missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylphenol, 98%

Product Code. 11487206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487206 25 g 25g
11374659 1 g 1g
11477206 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487206 Supplier Thermo Scientific Alfa Aesar Supplier No. B25527.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5306-98-9 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5306-98-9
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00060672
InChI Key KKFPXGXMSBBNJI-UHFFFAOYSA-N
Synonym 5-chloro-o-cresol, 5-chloro-2-cresol, o-cresol, 5-chloro, phenol, 5-chloro-2-methyl, 3-chloro-6-methylphenol, 2-methyl-5-chlorophenol, 4-chloro-2-hydroxytoluene, 5-chloro-2-methyl-phenol, phenol,5-chloro-2-methyl
PubChem CID 79192
IUPAC Name 5-chloro-2-methylphenol
SMILES CC1=CC=C(Cl)C=C1O

Specifications

Melting Point 70°C to 75°C
Boiling Point 109°C to 112°C (15 mmHg)
Quantity 25 g
UN Number UN3437
Beilstein 1906683
Formula Weight 142.59
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methylphenol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 5-Chloro-2-methylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5-Chloro-o-cresol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.