missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylaniline, 98%

Product Code. 10060730 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10060730 500 g Glass bottle 500g
10385880 5 g Glass bottle 5g
10395880 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10060730 Supplier Thermo Scientific Acros Supplier No. 109385000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-79-4 | C7H8ClN | 141.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-79-4
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007779
InChI Key WRZOMWDJOLIVQP-UHFFFAOYSA-N
Synonym 5-chloro-o-toluidine, 2-amino-4-chlorotoluene, 2-methyl-5-chloroaniline, fast red kb base, benzenamine, 5-chloro-2-methyl, ansibase red kb, red kb base, 4-chloro-2-aminotoluene, 3-chloro-6-methylaniline, pharmazoid red kb
PubChem CID 7260
ChEBI CHEBI:82422
IUPAC Name 5-chloro-2-methylaniline
SMILES CC1=CC=C(Cl)C=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 22°C
Color Yellow to Brown
Boiling Point 237°C
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(CH3)NH2
Refractive Index 1.5830 to 1.585
Quantity 500 g
Beilstein 12, 835
Solubility Information Solubility in water: < 1g/L (22°C). Other solubilities: soluble in alcohol,ether,benzene,chloroform and,dioxane
Formula Weight 141.6
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 5-Chloro-2-methylaniline, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Chloro-2-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.