missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxybenzoic Acid, TRC

Product Code. 30361409
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361409 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361409 Supplier Toronto Research Chemicals Supplier No. TRCC36791025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2-methoxybenzoic Acid
CAS 3438-16-2
Molecular Formula C8 H7 Cl O3
Quantity 25 g
Synonym 5-Chloro-2-methoxybenzoic Acid, Benzoic acid, 5-chloro-2-methoxy- (9CI, ACI), 5-Chloro-2-methoxybenzoic acid (ACI), o-Anisic acid, 5-chloro- (6CI, 7CI, 8CI), 2-Methoxy-5-chlorobenzoic acid, 5-Chloro-o-anisic acid
InChI Formula InChI=1S/C8H7ClO3/c1-12-7-3-2-5(9)4-6(7)8(10)11/h2-4H,1H3,(H,10,11)
SMILES COc1ccc(Cl)cc1C(=O)O
IUPAC Name 5-chloro-2-methoxybenzoic acid
Molecular Weight (g/mol) 186.59
Formula Weight 186.0084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.