missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxybenzenesulfonyl chloride, 98%

Product Code. 11466937
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466937 5 g 5g
11476937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466937 Supplier Thermo Scientific Alfa Aesar Supplier No. A14362.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22952-32-5 | C7H6Cl2O3S | 241.082 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22952-32-5
Molecular Formula C7H6Cl2O3S
Molecular Weight (g/mol) 241.082
MDL Number MFCD00052959
InChI Key FCJGLIMDVOTBLO-UHFFFAOYSA-N
Synonym 5-chloro-2-methoxybenzene-1-sulfonyl chloride, 5-chloro-2-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, 5-chloro-2-methoxy, chloro 5-chloro-2-methoxyphenyl sulfone, 5-chloro-2-methoxy-benzenesulfonyl chloride, acmc-1cq3l, ksc494k2b, 4-chloro-2-chlorosulphonyl anisole, 2-methoxy-5-chlorobenzenesulfonyl chloride, 5-chloro-2 methoxybenzenesulfonyl chloride
PubChem CID 2734272
IUPAC Name 5-chloro-2-methoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)Cl)S(=O)(=O)Cl

Specifications

Melting Point 100°C to 104°C
Quantity 5 g
UN Number UN3096
Formula Weight 241.1
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methoxybenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.