missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11341939
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11341939 5 g 5g
11351939 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341939 Supplier Thermo Scientific Alfa Aesar Supplier No. H26020.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 889939-48-4 | Molecular Formula: C8H8Cl2O3S | Molecular Weight: 255.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 889939-48-4
Molecular Formula C8H8Cl2O3S
Molecular Weight (g/mol) 255.109
MDL Number MFCD08445709
InChI Key BJRJHJZVSWZXIP-UHFFFAOYSA-N
Synonym 5-chloro-2-methoxy-4-methylbenzenesulfonylchloride, 5-chloro-2-methoxy-4-methylbenzene-1-sulfonyl chloride, pubchem5095, acmc-20am4m, 5-chloro-2-methoxy-4-methylphenylsulfonyl chloride, chloro 5-chloro-2-methoxy-4-methylphenyl sulfone, 5-chloranyl-2-methoxy-4-methyl-benzenesulfonyl chloride
PubChem CID 17389862
IUPAC Name 5-chloro-2-methoxy-4-methylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C=C1Cl)S(=O)(=O)Cl)OC

Specifications

Melting Point 126°C to 128°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 255.12
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.