missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-iodopyrimidine, 98%, Thermo Scientific™

Product Code. 15441279
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441279 250 mg 250mg
15421279 1 g 1g
15431279 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441279 Supplier Thermo Scientific Alfa Aesar Supplier No. H64185.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Useful Intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874676-81-0
Molecular Formula C4H2ClIN2
Molecular Weight (g/mol) 240.428
MDL Number MFCD08062390
InChI Key BWDLBDLGVMLSCS-UHFFFAOYSA-N
Synonym pyrimidine, 5-chloro-2-iodo, pubchem14957, acmc-209qnq, 5-chloro-2-iodo pyrimidine, ksc493q5d
PubChem CID 29938005
IUPAC Name 5-chloro-2-iodopyrimidine
SMILES C1=C(C=NC(=N1)I)Cl

Specifications

Color White
Quantity 250 mg
Beilstein 14516398
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 240.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Chloro-2-iodopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.