missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-hydroxy-3-methylbenzaldehyde, 95%, Thermo Scientific™

Product Code. 10098161
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10098161 1 g Amber Glass Bottle 1g
10110582 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10098161 Supplier Thermo Scientific Maybridge Supplier No. BTB03914DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 23602-63-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00275687
InChI Key NSKZAOKQZDLHGO-UHFFFAOYSA-N
Synonym benzaldehyde, 5-chloro-2-hydroxy-3-methyl, acmc-20am4p, 2-hydroxy-3-methyl-5-chlorobenzaldehyde, 5-chloro-2-hydroxy-3-methyl-benzaldehyde
PubChem CID 2774358
IUPAC Name 5-chloro-2-hydroxy-3-methylbenzaldehyde
SMILES CC1=C(C(=CC(=C1)Cl)C=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 170.59
Percent Purity 95%
Chemical Name or Material 5-Chloro-2-hydroxy-3-methylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.