missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorobenzaldehyde, 97%

Product Code. 11326589
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11326589

Brand: Thermo Scientific Alfa Aesar H26187.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96515-79-6 | C7H4ClFO | 158.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96515-79-6
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD03094518
InChI Key WDTUCEMLUHTMCB-UHFFFAOYSA-N
Synonym 2-fluoro-5-chlorobenzaldehyde, 3-chloro-6-fluoro benzaldehyde, 3-chloro-6-fluorobenzaldehyde, 5-chloro-2-fluoro-benzaldehyde, benzaldehyde, 5-chloro-2-fluoro, 2-fluoro-5-chloro-benzaldehyde, zlchem 333, pubchem1411, ksc486m0t, 2-fluoro-5-chloro benzaldehyde
PubChem CID 2773586
IUPAC Name 5-chloro-2-fluorobenzaldehyde
SMILES FC1=CC=C(Cl)C=C1C=O

Specifications

Melting Point 29°C to 32°C
Refractive Index 1.541
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 158.56
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluorobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.