missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2'-deoxyuridine, MP Biomedicals™

Product Code. 11431912 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431912 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11431912 Supplier MP Biomedicals Supplier No. 0210547891

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Readily incorporated, following phosphorylation, into newly synthesized DNA in place of thymidine. MP Biomedicals’ 5-chloro-2'-deoxyuridine (CldU) can be used as labeling substrate, thymidine analog, or to identify sites of DNA replication. Visible absorb

  • Used as a thymidine analog to study the miscoding potential of hypochlorous acid damage to DNA and DNA precursors
  • When used with antibody based immunofluorescent imaging, may be used in protocols to identify sites of DNA replication
  • May be used as a labeling substrate in conjunction with other halogenated uridine labeling substrates, such as bromodeoxyuridine (BrdU) and/or iododeoxyuridine (IdU)
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-90-8
Melting Point 166°C to 176°C
Molecular Formula C9H11ClN2O5
Quantity 1 g
Synonym 5-chloro-2'-deoxyuridine
Solubility Information Soluble in 1M NH4OH (20mg/1ml, colorless solution).
InChI Key NJCXGFKPQSFZIB-XVMARJQXSA-N
SMILES C1C(C(OC1N2C=C(C(=O)NC2=O)Cl)CO)O
IUPAC Name 5-chloro-1-[(2S,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 262.646
PubChem CID 13831089
Formula Weight 262.6
Physical Form Powder
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Chloro-2'-deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 5-Chloro-2'-deoxyuridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.