missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2'-deoxyuridine, MP Biomedicals™

Product Code. 11411912 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411912 0.1 g 100mg
11421912 0.25 g 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411912 Supplier MP Biomedicals Supplier No. 0210547880

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50-90-8
Molecular Formula C9H11ClN2O5
Molecular Weight (g/mol) 262.646
InChI Key NJCXGFKPQSFZIB-XVMARJQXSA-N
Synonym 5-chloro-2'-deoxyuridine
PubChem CID 13831089
IUPAC Name 5-chloro-1-[(2S,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1C(C(OC1N2C=C(C(=O)NC2=O)Cl)CO)O

Specifications

Color White
Assay Percent Range >98%
Quantity 0.1 g
Formula Weight 262.65
Physical Form Powder
Chemical Name or Material 5-Chloro-2'-deoxyuridine
compliance-icons compliance-icons
Product Identifier
  • 5-Chloro-2'-deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local, regional, national and international regulations.
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.