missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-[4-(dimethylamino)phenyl]benzimidazole, 95%, Thermo Scientific™

Product Code. 15419306
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1030574-44-7
MDL Number MFCD15173053
Synonym 4-(5-Chloro-2-benzimidazolyl)-N,N-dimethylaniline

Specifications

Quantity 250 mg
Beilstein 22393779
Formula Weight 271.75
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Chloro-2-[4-(dimethylamino)phenyl]benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'