Learn More
Alfa Aesar™ 5-Chloro-2-(4-chlorophenyl)benzimidazole, 95%
Brand: Alfa Aesar™ H64168.03
Code : NEW
Additional Details : CAS Number : 69498-30-2 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
5-Chloro-2-(4-chlorophenyl)benzimidazole | |
69498-30-2 | |
263.12 | |
MFCD11849216 |
Specifications
5-Chloro-2-(4-chlorophenyl)benzimidazole | |
69498-30-2 | |
95% | |
MFCD11849216 |
1g | |
Solid | |
263.12 |