missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10082224
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10082224 5 g Amber Glass Bottle 5g
10511112 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082224 Supplier Thermo Scientific Maybridge Supplier No. KM08674DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175203-78-8
Molecular Formula C6H2Cl2N2O3S
Molecular Weight (g/mol) 253.05
MDL Number MFCD00203878
InChI Key ZFBZICACMDZKNQ-UHFFFAOYSA-N
Synonym 5-chlorobenzofurazan-4-sulfonyl chloride, 5-chlorobenzo c 1,2,5 oxadiazole-4-sulfonyl chloride, pubchem5077, 2,1,3-benzoxadiazole-4-sulfonylchloride,5-chloro, 5-chloro-2,1,3-benzoxadiazole-4-sulphonyl chloride, 2,1,3-benzoxadiazole-4-sulfonylchloride, 5-chloro, 5-chloranyl-2,1,3-benzoxadiazole-4-sulfonyl chloride, 5-chloro-benzo 1,2,5 oxadiazole-4-sulfonyl chloride
PubChem CID 2779275
IUPAC Name 5-chloro-2,1,3-benzoxadiazole-4-sulfonyl chloride
SMILES ClC1=C(C2=NON=C2C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 253.06
Percent Purity 97%
Chemical Name or Material 5-chloro-2, 1, 3-benzoxadiazole-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.