missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-pentyne, 98%, ACROS Organics™

Brand:  Acros Organics 223420050

Code : 29

Additional Details : CAS Number : 14267-92-6 Weight : 0.05500kg Transport : UN number : 1993 Chem class : 3 Pack group : II


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10022880

  • £33.12 / 5g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

14267-92-6
102.56
UXFIKVWAAMKFQE-UHFFFAOYSA-N
84308
C#CCCCCl
C5H7Cl
MFCD00001014
5-chloro-1-pentyne, 1-pentyne, 5-chloro, 5-chloropentyne, 4-pentynyl chloride, 1-chloro-4-pentine, 1-chloro-4-pentyne, pent-4-ynyl chloride, 5-chloro-pent-1-yne, acmc-1bzfd, pent-4-yn-1-yl chloride
5-chloropent-1-yne

Specifications

5-Chloro-1-pentyne
15°C
Glass bottle
0.96
Colorless to Brown
98%
97.5
98%
Cl(CH2)3C∽CH
01, III, 958
UXFIKVWAAMKFQE-UHFFFAOYSA-N
5-chloropent-1-yne
84308
Liquid
0.9600g/mL
Authentic
1.4430 to 1.445
67°C to 69°C (145.0mmHg)
5g
14267-92-6
100.0
C5H7Cl
MFCD00001014
5-chloro-1-pentyne, 1-pentyne, 5-chloro, 5-chloropentyne, 4-pentynyl chloride, 1-chloro-4-pentine, 1-chloro-4-pentyne, pent-4-ynyl chloride, 5-chloro-pent-1-yne, acmc-1bzfd, pent-4-yn-1-yl chloride
C#CCCCCl
102.56
102.56
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Chloro-1-pentyne

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents