missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-1-methylimidazole, TRC
Click to view available options
Quantity:
2 g
Unit Size:
2g
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-1-methylimidazole |
| CAS | 872-49-1 |
| Molecular Formula | C4 H5 Cl N2 |
| Quantity | 2 g |
| Synonym | Azathioprine Impurity 9 |
| InChI Formula | InChI=1S/C4H5ClN2/c1-7-3-6-2-4(7)5/h2-3H,1H3 |
| SMILES | Cn1cncc1Cl |
| IUPAC Name | 5-chloro-1-methylimidazole |
| Molecular Weight (g/mol) | 116.55 |
| Formula Weight | 116.0141 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction