missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methylimidazole, 98%, Thermo Scientific™

Product Code. 11392697
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11392697

Brand: Thermo Scientific Alfa Aesar L11899.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-49-1
Molecular Formula C4H5ClN2
Molecular Weight (g/mol) 116.548
MDL Number MFCD00014505
InChI Key NYDGOZPYEABERA-UHFFFAOYSA-N
Synonym 5-chloro-1-methyl-1h-imidazole, 1-methyl-5-chloroimidazole, 1h-imidazole, 5-chloro-1-methyl, 5-chloro-1-methylimidazole, zlchem 661, pubchem7612, acmc-209qj7, 5-chloro-1-methyl imidazole, 5-chloro-1-methyl-imidazole, ksc494e7d
PubChem CID 70105
IUPAC Name 5-chloro-1-methylimidazole
SMILES CN1C=NC=C1Cl

Specifications

Density 1.25
Boiling Point 89°C to 90°C (14 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.511
Quantity 250 mg
Beilstein 110509
Formula Weight 116.55
Percent Purity 98%
Chemical Name or Material 5-Chloro-1-methylimidazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.