missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-indanone, 99%

Product Code. 11423867
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11423867

Brand: Thermo Scientific Alfa Aesar A18308.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 42348-86-7 | C9H7ClO | 166.60 g/mol

5-Chloro-1-indanone may be used as starting reagent for the preparation of 5-chloro-2-methoxycarbonyl-1-indanone. It may also be used for the preparation of important biomedical compounds such as anticonvulsants, anticholinergics and diarylsulfonylureas, having potential activity against solid tumors. It participates in the Irie?s synthesis of substituted pyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-1-indanone may be used as starting reagent for the preparation of 5-chloro-2-methoxycarbonyl-1-indanone. It may also be used for the preparation of important biomedical compounds such as anticonvulsants, anticholinergics and diarylsulfonylureas, having potential activity against solid tumors. It participates in the Irie′s synthesis of substituted pyridines.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42348-86-7
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.60
MDL Number MFCD00041456
InChI Key MEDSHTHCZIOVPU-UHFFFAOYSA-N
Synonym 5-chloro-1-indanone, 5-chloro-2,3-dihydro-1h-inden-1-one, 5-chloroindan-1-one, 5-chloro-indan-1-one, 1h-inden-1-one, 5-chloro-2,3-dihydro, 5-chloroindanone, 5-chloroindone, pubchem2250, 1-indanone, 5-chloro, 5-chloro indanone
PubChem CID 142599
IUPAC Name 5-chloro-2,3-dihydro-1H-inden-1-one
SMILES ClC1=CC=C2C(=O)CCC2=C1

Specifications

Melting Point 92°C to 96°C
Boiling Point 124°C to 125°C (3 mmHg)
Quantity 5 g
Beilstein 1448000
Solubility Information Insoluble in water.
Formula Weight 166.61
Percent Purity 99%
Chemical Name or Material 5-Chloro-1-indanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.