missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-1-indanone, 98%, ACROS Organics™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 93°C to 98°C |
| Color | White to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Quantity | 250 mg |
| Formula Weight | 166.61 |
| Percent Purity | ≥97.5% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction