missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,3-dimethylpyrazole, 98+%, Thermo Scientific™

Catalog Number p-86869
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
54454-10-3
C5H7ClN2
130.58
MFCD00051650
DDUSLFAWARYAPR-UHFFFAOYSA-N
521507
5-chloro-1,3-dimethyl-1H-pyrazole
CN1N=C(C)C=C1Cl
This item is not returnable. View return policy
54454-10-3
C5H7ClN2
130.58
MFCD00051650
DDUSLFAWARYAPR-UHFFFAOYSA-N
521507
5-chloro-1,3-dimethyl-1H-pyrazole
CN1N=C(C)C=C1Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
Quantity 1 g
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole, 5-chloro-1,3-dimethyl, 1,3-dimethyl-5-chloropyrazole, pubchem9980, acmc-20am54, 5-chloro-1,3-dimethyl-pyrazole, 5-chloranyl-1,3-dimethyl-pyrazole, 1,3-dimethyl-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-1,3-dimethyl, 5-chloro-1,3-dimethyl-1h-pyrazole #
Formula Weight 130.58
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 5-Chloro-1, 3-dimethylpyrazole, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Chloro-1,3-dimethylpyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'