missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-1,3-dimethyl-1H-pyrazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10039111
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10039111 10 g Amber Glass Bottle 10g
10651673 1 g Amber Glass Bottle 1g
10798435 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10039111 Supplier Thermo Scientific Maybridge Supplier No. CD10088EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 88398-93-0
Molecular Formula C5H6Cl2N2O2S
Molecular Weight (g/mol) 229.075
MDL Number MFCD00052536
InChI Key HWGVUNSKAPCFNF-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole-4-sulfonyl chloride, 5-chloro-1,3-dimethyl-4-pyrazolesulfonyl chloride, 5-chloro-1,3-dimethyl-1h-pyrazole-4-sulphonyl chloride, 5-chloro-1,3-dimethyl-pyrazole-4-sulfonyl chloride, 5-chloro-1,3-dimethylpyrazole-4-sulfonylchloride, 5-chloranyl-1,3-dimethyl-pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride, 5-chloro-1,3-dimethyl, 5-chloro-1,3-dimethyl-1h-pyrazol-4yl-sulfonyl chloride, 5-chloro-1,3-dimethyl-1h-pyrazol-4-yl sulfonyl chloride, 5-chloro-1,3-dimethyl-1h-pyrazol-4-ylsulphonyl chloride
PubChem CID 2777126
IUPAC Name 5-chloro-1,3-dimethylpyrazole-4-sulfonyl chloride
SMILES CC1=NN(C(=C1S(=O)(=O)Cl)Cl)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 229.09
Percent Purity 97%
Chemical Name or Material 5-chloro-1, 3-dimethyl-1H-pyrazole-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.