missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-1,3-dimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10764193
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 27006-82-2
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.584
MDL Number MFCD00232043
InChI Key RRWQERXMLIEDKJ-UHFFFAOYSA-N
PubChem CID 2779652
IUPAC Name 5-chloro-1,3-dimethylpyrazole-4-carboxylic acid
SMILES CC1=NN(C(=C1C(=O)O)Cl)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole-4-carboxylic acid, 5-chloro-1,3-dimethyl-4-pyrazolecarboxylic acid, pubchem23627, chlorodimethylpyrazolecarboxylicacid, 4-carboxy-5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid,5-chloro-1,3-dimethyl, 1h-pyrazole-4-carboxylic acid, 5-chloro-1,3-dimethyl, 1h-pyrazole-4-carboxylicacid, 5-chloro-1,3-dimethyl
Formula Weight 174.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-1, 3-dimethyl-1H-pyrazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'